2,3,4,5,6-Pentafluorobenzyl alcohol ≥98%

Supplier: BeanTown Chemical

Synonyms: (Perfluorophenyl)methanol, (2,3,4,5,6-Pentafluorophenyl)methanol

124440-100G 124440-25G 124440-5G
BT124440-100GEA 575 USD
BT124440-100G BT124440-25G BT124440-5G
2,3,4,5,6-Pentafluorobenzyl alcohol ≥98%
2,3,4,5,6-Pentafluorobenzyl alcohol

CAS: 440-60-8; EC No: 207-126-7; MDL No: MFCD00004602; RTECS: DP0695000 Solid; Molecular Formula: C7H3F5O; MW: 198.09 Melting Point: 37-38°; Boiling Point: 114-115°/60 mmHg; Flash point: 88°C (190°F)


  • RTECS: DP0695000
  • Solid

Formula: C₇H₃F₅O
MW: 198.09 g/mol
Boiling Pt: 114…115 °C/60 mmHg
Melting Pt: 37…38 °C
Flash Pt: 88 °C (190 °F)
MDL Number: MFCD00004602
CAS Number: 440-60-8
EINECS: 207-126-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR