trans-1-Ethoxy-4-(4-n-propycyclohexyl)benzene ≥97%
Supplier: BeanTown Chemical
Synonyms:
1-Ethoxy-4-(trans-4-propylcyclohexyl)benzene
124425-1G
124425-5G
BT124425-1GEA
74
USD
BT124425-1G
BT124425-5G
trans-1-Ethoxy-4-(4-n-propycyclohexyl)benzene ≥97%
trans-1-Ethoxy-4-(4-n-propycyclohexyl)benzene
CAS: 80944-44-1; EC No: 279-633-1; MDL No: MFCD06658195
Solid; Molecular Formula: C17H26O; MW: 246.39
Melting Point: 40-42°
- Solid
Formula:
C₁₇H₂₆O MW: 246.394 g/mol Melting Pt: 40…42 °C |
MDL Number:
MFCD06658195 CAS Number: 80944-44-1 EINECS: 279-633-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...