4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulfonylamino)-5-pyrimidinecarboxaldehyde ≥99%
Supplier: BeanTown Chemical
Synonyms:
N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulphonamide, N-[4-(4-Fluorophenyl)-5-formyl-6-(1-methylethyl)-2-pyrimidinyl]-N-methyl-methanesulphonamide, N-(4-(4-Fluorophenyl)-5-formyl-6-isopropylpyrimidin-2-yl)-N-methylmethanesulphonamide
124410-100G
124410-25G
124410-5G
BT124410-100GEA
351
USD
BT124410-100G
BT124410-25G
BT124410-5G
4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulfonylamino)-5-pyrimidinecarboxaldehyde ≥99%
4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulfonylamino)-5-pyrimidinecarboxaldehyde
CAS: 147118-37-4; MDL No: MFCD08458342
Solid; Molecular Formula: C16H18FN3O3S
; MW: 351.40
- Solid
Formula:
C₁₆H₁₈FN₃O₃S MW: 351.40 g/mol |
MDL Number:
MFCD08458342 CAS Number: 147118-37-4 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...