(1S,2R)-(-)-10,2-Camphorsultam ≥99%

Supplier: BeanTown Chemical
Warning

Synonyms: (-)-10,2-Camphorsultam, (2R)-(-)-Bornane-10,2-sultam, (1S,5R)-10,10-Dimethyl-3-thia-4-azatricyclo[5.2.1.0(1,5)]decane 3,3-dioxide, (1S)-(−)-2,10-Camphorsultam, [3aS-(3aalpha,6alpha,7abeta)]-Hexahydro-8,8-dimethyl-2,2-dioxide-3H-3a,6-methano-2,1-benzisothiazole, (3aS,6R,7aR)-8,8-Dimethylhexahydro-1H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide, (3aS,6S,7aS)-8,8-Dimethylhexahydro-1H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide, (1S,2R,4R)-2,10-camphorsultam, (-)-exo-10,2-Bornanesultam, (2R)-Bornane-10,2-sultam

124355-1G 124355-25G 124355-5G
BT124355-1GEA 80 USD
BT124355-1G BT124355-25G BT124355-5G
(1S,2R)-(-)-10,2-Camphorsultam ≥99%
(1S,2R)-(-)-10,2-Camphorsultam

CAS: 94594-90-8; MDL No: MFCD00066271 Crystalline; Molecular Formula: C10H17NO2S ; MW: 215.31 Melting Point: 181-183° Optical Rotation: [α]19/D −32°, c = 5 in chloroform


  • Optical Rotation: [α]19/D −32°, c = 5 in chloroform

Formula: C₁₀H₁₇NO₂S
MW: 215.32 g/mol
Melting Pt: 181…183 °C
MDL Number: MFCD00066271
CAS Number: 94594-90-8

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR