Aluminium oxide, γ-α-phase ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
124345-100G
124345-25G
124345-500G
BT124345-100GEA
116
USD
BT124345-100G
BT124345-25G
BT124345-500G
Aluminium oxide, γ-α-phase ≥99.99% (trace metals basis)
Aluminium oxide, γ-α-phase
CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424
0.3 Micron Powder, S.A. 15 m2/g; Molecular Formula: Al2O3; MW: 101.96
Melting Point: 2040°
- 0.3 Micron Powder, S.A. 15 m²/g
Formula:
Al₂O₃ Melting Pt: 2040 °C |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...