Cobalt(II) hydroxide ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
124340-100G
124340-2KG
124340-500G
BT124340-100GEA
47.78
USD
BT124340-100G
BT124340-2KG
BT124340-500G
Cobalt(II) hydroxide ≥99.9% (trace metals basis)
Cobalt(II) hydroxide
CAS: 21041-93-0; EC No: 244-166-4; MDL No: MFCD00016015; RTECS: GG2800000
Powder; Linear Formula: Co(OH)2; MW: 74.93
Density (g/mL): 3.597
Air Sensitive
- RTECS: GG2800000
- Air Sensitive
- Powder
Formula:
H₂CoO₂ Density: 3.597 g/mL |
MDL Number:
MFCD00016015 CAS Number: 21041-93-0 EINECS: 244-166-4 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...