4-Chlorophenylhydrazinium chloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Chlorophenylhydrazine hydrochloride, 1-(4-Chlorophenyl)hydrazine hydrochloride
124305-100G
124305-25G
124305-5G
BT124305-100GEA
336
USD
BT124305-100G
BT124305-25G
BT124305-5G
4-Chlorophenylhydrazinium chloride ≥97%
4-Chlorophenylhydrazinium chloride
CAS: 1073-70-7; EC No: 214-030-9; MDL No: MFCD00012943
Powder; Molecular Formula: C6H7ClN2·HCl; MW: 179.05
Melting Point: 216° (decomposes)
- Powder
Formula:
C₆H₈Cl₂N₂ MW: 179.05 g/mol Melting Pt: 216 °C (dec.) |
MDL Number:
MFCD00012943 CAS Number: 1073-70-7 EINECS: 214-030-9 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...