2-Hydroxy-5-nitropyridine ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Nitro-2-pyridinol

124290-100G 124290-25G
BT124290-100GEA 178 USD
BT124290-100G BT124290-25G
2-Hydroxy-5-nitropyridine ≥98%
2-Hydroxy-5-nitropyridine

CAS: 5418-51-9; EC No: 226-525-7; MDL No: MFCD00006276 Crystalline/Powder; Molecular Formula: C5H4N2O3 ; MW: 140.10 Melting Point: 188-191°


  • Crystalline/Powder

Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
Melting Pt: 188…191 °C
MDL Number: MFCD00006276
CAS Number: 5418-51-9
EINECS: 226-525-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR