Behenic acid ≥85%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Docosanoic acid

124270-1KG 124270-250G 124270-50G
BT124270-1KGEA 116.95 USD
BT124270-1KG BT124270-250G BT124270-50G
Behenic acid ≥85%, Technical Grade
Behenic acid

CAS: 112-85-6; EC No: 204-010-8; MDL No: MFCD00002807 Solid; Linear Formula: CH3(CH2)20CO2H; Molecular Formula: C22H44O2; MW: 340.58 Melting Point: 72-80°


  • Solid

Formula: C₂₂H₄₄O₂
MW: 340.59 g/mol
Melting Pt: 72…80 °C
MDL Number: MFCD00002807
CAS Number: 112-85-6
EINECS: 204-010-8

Order Now

Specification Test Results

Assay 85%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR