1,4,8,11-Tetraazacyclotetradecane ≥98%
Supplier: BeanTown Chemical
124250-1G
124250-5G
BT124250-1GEA
67
USD
BT124250-1G
BT124250-5G
1,4,8,11-Tetraazacyclotetradecane ≥98%
1,4,8,11-Tetraazacyclotetradecane
CAS: 295-37-4; EC No: 206-039-1; MDL No: MFCD00005105; RTECS: XA5254800
Powder; Molecular Formula: C10H24N4; MW: 200.32
Melting Point: 184-186°
Air Sensitive
- RTECS: XA5254800
- Air Sensitive
- Powder
Formula:
C₁₀H₂₄N₄ MW: 200.33 g/mol Melting Pt: 184…186 °C |
MDL Number:
MFCD00005105 CAS Number: 295-37-4 EINECS: 206-039-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...