Casein

Supplier: BeanTown Chemical

124240-10KG 124240-2.5KG 124240-500G
BT124240-10KGEA 507 USD
BT124240-10KG BT124240-2.5KG BT124240-500G
Casein
Proteins and Peptides

EC No: 232-555-1
RTECS: FI3519500
Melting Point: 280 °C (decomposes)
Sensitivity: Hygroscopic
Merck Number: 14,1883


Información de suministro: Non-hazardous for shipping.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR