L(+)-Potassium hydrogen tartrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Potassium hydrogen L-tartrate, Potassium acid tartrate, Potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Potassium Bitartrate, (R,R)-(+)-Tartaric acid potassium salt, L(+)-Tartaric acid potassium salt

124215-1KG 124215-250G 124215-5KG
BT124215-1KGEA 103.24 USD
BT124215-1KG BT124215-250G BT124215-5KG
L(+)-Potassium hydrogen tartrate ≥98%
L(+)-Potassium hydrogen tartrate

CAS: 868-14-4; EC No: 212-769-1; MDL No: MFCD00065392; RTECS: WW8223000 Powder; Linear Formula: KOOCCH(OH)CH(OH)COOH; Molecular Formula: C4H5KO6; MW: 188.18 Melting Point: 250° Density (g/mL): 1.954; Optical Rotation: [α]20/D +10°, c = 2 in 10% HCl


  • RTECS: WW8223000
  • Optical rotation: [α]20/D +10°, c = 2 in 10% HCl
  • Powder

Formula: KOOC(CHOH)₂COOH
MW: 188.18 g/mol
Melting Pt: 250
Density: 1.954 g/mL
MDL Number: MFCD00065392
CAS Number: 868-14-4
EINECS: 212-769-1
Merck Index: 13,07697
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=355f72f319f5589f12d60c6dfffecdec31cee083&vid=4a750ad5d4a91fd4c2885832ee80b7b64cdcc42a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR