(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥97%

Supplier: BeanTown Chemical

Synonyms: (S)-(-)-BINAP, (S)-BINAP, (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (S)-(-)-2,2'-Bis(diphenylphoshino)-1,1'-binaphthyl

124195-1G 124195-250MG 124195-5G
BT124195-1GEA 104 USD
BT124195-1G BT124195-250MG BT124195-5G
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥97%
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

CAS: 76189-56-5; MDL No: MFCD00010805 Crystalline/Powder; Molecular Formula: C44H32P2; MW: 622.67 Melting Point: 238-240° Optical Rotation: [α]20/D -222°, c = 0.5 in benzene Air Sensitive


  • Optical rotation: [α]20/D -222°, c = 0.5 in benzene
  • Air Sensitive
  • Crystalline/Powder

Formula: C₄₄H₃₂P₂
MW: 622.69 g/mol
Melting Pt: 238…240 °C
MDL Number: MFCD00010805
CAS Number: 76189-56-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR