5-Nitrouracil ≥98%
Supplier: BeanTown Chemical
Synonyms:
Nitrouracil, 5-Nitropyrimidine-2,4(1H,3H)-dione, 2,4-Dihydroxy-5-nitropyrimidine
124185-100G
124185-25G
124185-500G
BT124185-100GEA
122
USD
BT124185-100G
BT124185-25G
BT124185-500G
5-Nitrouracil ≥98%
5-Nitrouracil
CAS: 611-08-5; EC No: 210-250-4; MDL No: MFCD00006021; RTECS: UV9104545
Powder; Molecular Formula: C4H3N3O4; MW: 157.08
Melting Point:
- RTECS: UV9104545
- Powder
Formula:
C₄H₃N₃O₄ MW: 157.09 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00006021 CAS Number: 611-08-5 EINECS: 210-250-4 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...