2-Chloro-5-nitropyridine ≥99%
Supplier: BeanTown Chemical
124180-100G
124180-25G
BT124180-100GEA
433
USD
BT124180-100G
BT124180-25G
2-Chloro-5-nitropyridine ≥99%
2-Chloro-5-nitropyridine
CAS: 4548-45-2; EC No: 224-908-3; MDL No: MFCD00006240; RTECS: US7175000
Powder; Molecular Formula: C5H3ClN2O2; MW: 158.54
Melting Point: 105-108°
- RTECS: US7175000
- Powder
Formula:
C₅H₃ClN₂O₂ MW: 158.54 g/mol Melting Pt: 107 |
MDL Number:
MFCD00006240 CAS Number: 4548-45-2 EINECS: 224-908-3 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...