Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine
Supplier: BeanTown Chemical
Synonyms:
((2,4,6-Tri-isopropyl)phenyl)dicyclohexylphosphine
124165-500MG
124165-5G
BT124165-500MGEA
63
USD
BT124165-500MG
BT124165-5G
Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine
Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine
CAS: 303111-96-8; MDL No: MFCD11656073
Solid; Molecular Formula: C27H45P ; MW: 400.62
Melting Point: 138-140°
Air Sensitive
- Air Sensitive
- Solid
Formula:
C₂₇H₄₅P Melting Pt: 138…140 °C |
MDL Number:
MFCD11656073 CAS Number: 303111-96-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...