2,3-Diaminonaphthalene ≥97%

Supplier: BeanTown Chemical

Synonyms: 2,3-Naphthalenediamine, DAN, 2,3-naphthylenediamine

124150-1G 124150-5G
BT124150-1GEA 124.99 USD
BT124150-1G BT124150-5G
2,3-Diaminonaphthalene ≥97%
2,3-Diaminonaphthalene

CAS: 771-97-1; EC No: 212-241-0; MDL No: MFCD00004116 Powder; Molecular Formula: C10H10N2; MW: 158.20


  • Powder

Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
MDL Number: MFCD00004116
CAS Number: 771-97-1
EINECS: 212-241-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR