Magnesium aluminate ≥99% (trace metals basis)

Supplier: BeanTown Chemical

124125-100G 124125-500G
BT124125-100GEA 46 USD
BT124125-100G BT124125-500G
Magnesium aluminate ≥99% (trace metals basis)
Magnesium aluminate

CAS: 12068-51-8; EC No: 235-100-5; MDL No: MFCD00049475 -325 Mesh Powder; Molecular Formula: Al2MgO4; MW: 142.26


  • -325 Mesh Powder

Formula: Al₂MgO₄
MDL Number: MFCD00049475
CAS Number: 12068-51-8
EINECS: 235-100-5

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR