Cobalt(II) bromide hydrate
Supplier: BeanTown Chemical
Synonyms:
Cobalt dibromide hydrate, Cobaltous bromide hydrate
124010-250G
124010-50G
BT124010-250GEA
316
USD
BT124010-250G
BT124010-50G
Cobalt(II) bromide hydrate
Cobalt(II) bromide hydrate
CAS: 85017-77-2; EC No: 232-166-7; MDL No: MFCD00661152; RTECS: GF9595000
Crystalline/Granular; Linear Formula: CoBr2·xH2O; MW: 218.75 (anhydrous)
Melting Point: 47-48° (decomposes)
Hygroscopic
- RTECS: GF9595000
- Hygroscopic
- Granular
Formula:
Br₂Co·H₂O MW: 236.76 g/mol Melting Pt: 47…48 °C (dec.) |
MDL Number:
MFCD00661152 CAS Number: 85017-77-2 EINECS: 232-166-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...