Amantadine hydrochloride ≥99%

Supplier: BeanTown Chemical

Synonyms: 1-Adamantylamine hydrochloride, 1-Aminotricyclo[3.3.1.1(3,7)]decane hydrochloride, 1-Aminotricyclo(3.3.1.1(3,7))decan HCl, 1-Aminoadamantane hydrochloride, 1-Adamantanammonium chloride, 1-Adamantanamine hydrochloride, Tricyclo[3.3.1.1(3,7)]dec-1-ylamine hydrochloride, Adamantan-1-amine hydrochloride, 1-Adamantaneammonium Chloride

123995-100G 123995-25G 123995-500G
BT123995-100GEA 178 USD
BT123995-100G BT123995-25G BT123995-500G
Amantadine hydrochloride ≥99%
Amantadine hydrochloride

CAS: 665-66-7; EC No: 211-560-2; MDL No: MFCD00074723; RTECS: AU4375000 Crystalline/Solid; Molecular Formula: C10H17N·HCl; MW: 187.71 Hygroscopic


  • RTECS: AU4375000
  • Hygroscopic
  • Crystalline/Solid

Formula: C₁₀H₁₈ClN
MDL Number: MFCD00074723
CAS Number: 665-66-7
EINECS: 211-560-2

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR