Zinc ≥99.9999% (trace metals basis), rods Ø 8 mm
Supplier: BeanTown Chemical
123990-250MM
123990-50MM
BT123990-250MMEA
2008
USD
BT123990-250MM
BT123990-50MM
Zinc ≥99.9999% (trace metals basis), rods Ø 8 mm
Zinc
CAS: 7440-66-6; EC No: 231-175-3; MDL No: MFCD00011291
Solid; Molecular Formula: Zn; MW: 65.39
Melting Point: 420°; Boiling Point: 907°
Density (g/mL): 7.133
- Solid
Formula:
Zn Boiling Pt: 907 °C Melting Pt: 420 °C Density: 7.133 g/mL |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 |
Specification Test Results
Assay | 99.9999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...