3,4,5,6-Tetrachlorofluorescein
Supplier: BeanTown Chemical
123975-1G
123975-5G
BT123975-1GEA
30
USD
BT123975-1G
BT123975-5G
3,4,5,6-Tetrachlorofluorescein
3,4,5,6-Tetrachlorofluorescein
CAS: 6262-21-1; EC No: 228-413-3; MDL No: MFCD00010030
Powder; Molecular Formula: C20H8Cl4O5; MW: 470.10
Melting Point: 304° (decomposes)
- Powder
Formula:
C₂₀H₈Cl₄O₅ Melting Pt: 304 °C (dec.) |
MDL Number:
MFCD00010030 CAS Number: 6262-21-1 EINECS: 228-413-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...