Tetramethylthiuram monosulphide ≥97%
Supplier: BeanTown Chemical
Synonyms:
Bis(dimethylthiocarbamyl) sulphide
123955-100G
123955-25G
123955-500G
BT123955-100GEA
64
USD
BT123955-100G
BT123955-25G
BT123955-500G
Tetramethylthiuram monosulphide ≥97%
Tetramethylthiuram monosulphide
CAS: 97-74-5; EC No: 202-605-7; MDL No: MFCD00014870; RTECS: WQ1750000
Powder; Linear Formula: [(CH3)2NCS]2S; MW: 208.37
Melting Point: 107-111°; Flash point: 156°C (313°F)
Density (g/mL): 1.37
- RTECS: WQ1750000
- Powder
Formula:
C₆H₁₂N₂S₃ MW: 208.37 g/mol Melting Pt: 110 Density: 1.37 g/mL Flash Pt: 156 °C (313 °F) |
MDL Number:
MFCD00014870 CAS Number: 97-74-5 EINECS: 202-605-7 UN: 3077 ADR: 9 III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...