4,4'-Azobis(4-cyano-1 pentanol)
Supplier: BeanTown Chemical
123920-1G
123920-5G
BT123920-1GEA
155
USD
BT123920-1G
BT123920-5G
4,4'-Azobis(4-cyano-1 pentanol)
4,4'-Azobis(4-cyano-1 pentanol)
CAS: 4693-47-4
Solid; Molecular Formula: C12H20N4O2; MW: 252.31
- Solid
Formula:
C₁₂H₂₀N₄O₂ |
CAS Number:
4693-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...