Butyl (±)-lactate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Butyl 2-hydroxypropanoate, n-Butyl lactate, Butyl (±)-2-hydroxypropionate, (±)-Butyl lactate
123900-2KG
123900-500G
BT123900-2KGEA
325
USD
BT123900-2KG
BT123900-500G
Butyl (±)-lactate ≥98%
Butyl (±)-lactate
CAS: 138-22-7; EC No: 205-316-4; MDL No: MFCD00004519; RTECS: OD4025000
Liquid; Linear Formula: CH3CH(OH)COO(CH2)3CH3; Molecular Formula: C7H14O3; MW: 146.18
Melting Point: -28°; Boiling Point: 185-187°; Flash point: 69°C (156°F)
Density (g/mL): 0.984; Refractive Index: 1.421
- RTECS: OD4025000
- Refractive index: 1.421
- Liquid
Formula:
C₇H₁₄O₃ MW: 146.19 g/mol Boiling Pt: 185…187 °C Melting Pt: –28 °C Density: 0.984 g/mL Flash Pt: 69 °C (156 °F) |
MDL Number:
MFCD00004519 CAS Number: 138-22-7 EINECS: 205-316-4 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...