Tetrapropylammonium chloride ≥99%
Supplier: BeanTown Chemical
123890-1G
123890-25G
123890-5G
BT123890-1GEA
23
USD
BT123890-1G
BT123890-25G
BT123890-5G
Tetrapropylammonium chloride ≥99%
Tetrapropylammonium chloride
CAS: 5810-42-4; EC No: 227-375-5; MDL No: MFCD00038729
Crystalline; Linear Formula: (CH3CH2CH2)4N(Cl); Molecular Formula: C12H28ClN; MW: 221.81
Melting Point: 240-242°
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
C₁₂H₂₈ClN Melting Pt: 240…242 °C |
MDL Number:
MFCD00038729 CAS Number: 5810-42-4 EINECS: 227-375-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...