4-Aminobenzyl alcohol ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-(Hydroxymethyl)aniline, (4-Aminophenyl)methanol
123865-25G
123865-5G
BT123865-25GEA
388
USD
BT123865-25G
BT123865-5G
4-Aminobenzyl alcohol ≥98%
4-Aminobenzyl alcohol
CAS: 623-04-1; EC No: 210-767-5; MDL No: MFCD00014782
Crystalline; Molecular Formula: C7H9NO; MW: 123.15
Melting Point: 60-65°
Air Sensitive, Light Sensitive
- Air Sensitive, Light Sensitive
- Crystalline
Formula:
C₇H₉NO MW: 123.15 g/mol Melting Pt: 60…65 °C |
MDL Number:
MFCD00014782 CAS Number: 623-04-1 EINECS: 210-767-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...