3-Hydroxybenzaldehyde ≥97%
Supplier: BeanTown Chemical
123860-100G
123860-25G
123860-500G
BT123860-100GEA
50
USD
BT123860-100G
BT123860-25G
BT123860-500G
3-Hydroxybenzaldehyde ≥97%
3-Hydroxybenzaldehyde
CAS: 100-83-4; EC No: 202-892-9; MDL No: MFCD00003368; RTECS: CU6450000
Solid; Molecular Formula: C7H6O2; MW: 122.12
Melting Point: 100-103°; Boiling Point: 191°/50 mmHg
Air Sensitive
- RTECS: CU6450000
- Air Sensitive
- Solid
Formula:
C₇H₆O₂ MW: 122.12 g/mol Boiling Pt: 191 °C/50 mmHg Melting Pt: 100…103 °C |
MDL Number:
MFCD00003368 CAS Number: 100-83-4 EINECS: 202-892-9 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...