Lanthanum(III) 2,4-pentanedionate hydrate

Supplier: BeanTown Chemical

Synonyms: Lanthanum(III) acetylacetonate hydrate, Tris(2,4-pentanedionato)lanthanum(III) hydrate, Acetylacetone Lanthanum(III) hydrate

123845-10G 123845-50G
BT123845-10GEA 49 USD
BT123845-10G BT123845-50G
Lanthanum(III) 2,4-pentanedionate hydrate
Lanthanum(III) 2,4-pentanedionate hydrate

CAS: 64424-12-0; EC No: 238-187-8; MDL No: MFCD00149057 Powder; Molecular Formula: C15H21LaO6·xH2O ; MW: 454.24 (anhydrous) Hygroscopic


  • Hygroscopic
  • Powder

Formula: C₁₅H₂₁LaO₆·xH₂O
MW: 436.24 g/mol (anhy)
MDL Number: MFCD00149057
CAS Number: 64424-12-0
EINECS: 238-187-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR