Tetrahexylammonium bromide ≥98%

Supplier: BeanTown Chemical

123840-10G 123840-250G 123840-50G
BT123840-10GEA 56.54 USD
BT123840-10G BT123840-250G BT123840-50G
Tetrahexylammonium bromide ≥98%
Tetrahexylammonium bromide

CAS: 4328-13-6; EC No: 224-363-1; MDL No: MFCD00011858 Crystalline; Linear Formula: [CH3(CH2)5]4N(Br); Molecular Formula: C24H52BrN; MW: 434.58 Melting Point: 99-100° Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₂₄H₅₂BrN
MW: 434.59 g/mol
Melting Pt: 104
MDL Number: MFCD00011858
CAS Number: 4328-13-6
EINECS: 224-363-1

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR