(-)-Quinine hydrochloride dihydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Quinine hydrochloride dihydrate, Quinine monohydrochloride dihydrate, (R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol hydrochloride dihydrate
123830-10G
123830-50G
BT123830-10GEA
50.68
USD
BT123830-10G
BT123830-50G
(-)-Quinine hydrochloride dihydrate ≥99%
(-)-Quinine hydrochloride dihydrate
CAS: 6119-47-7; EC No: 231-437-7; MDL No: MFCD00151248; RTECS: VA7700000
Solid; Molecular Formula: C20H24N2O2·HCl·2H2O; MW: 396.91
Light Sensitive
- RTECS: VA7700000
- Light Sensitive
- Solid
Formula:
C₂₀H₂₅ClN₂O₂·2H₂O |
MDL Number:
MFCD00151248 CAS Number: 6119-47-7 EINECS: 231-437-7 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...