N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide
Supplier: BeanTown Chemical
Synonyms:
N-(2,6-Dimethylphenyl)-1-piperazineacetamide
123795-1G
123795-250MG
123795-5G
BT123795-1GEA
111
USD
BT123795-1G
BT123795-250MG
BT123795-5G
N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide
N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide
CAS: 5294-61-1; MDL No: MFCD06384973
Solid; Molecular Formula: C14H21N3O; MW: 247.34
Melting Point: 110-112°
- Solid
Formula:
C₁₄H₂₁N₃O MW: 247.34 Melting Pt: 110…112 °C |
MDL Number:
MFCD06384973 CAS Number: 5294-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...