N-Fmoc-L-valine ≥98%

Supplier: BeanTown Chemical

123780-25G 123780-5G
BT123780-25GEA 163 USD
BT123780-25G BT123780-5G
N-Fmoc-L-valine ≥98%
N-Fmoc-L-valine

CAS: 68858-20-8; EC No: 272-515-0; MDL No: MFCD00037124 Solid; Molecular Formula: C20H21NO4 ; MW: 339.39 Melting Point: 143-145° Optical Rotation: [α]20/D -17±1°, c = 1% in DMF


  • Optical rotation: [α]20/D -17±1°, c = 1% in DMF
  • Solid

Formula: C₂₀H₂₁NO₄
MW: 339.4 g/mol
Melting Pt: 143…145 °C
MDL Number: MFCD00037124
CAS Number: 68858-20-8
EINECS: 272-515-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR