Tannic acid ACS
Supplier: BeanTown Chemical
Synonyms:
Gallotannin, Tannin
123775-100G
123775-500G
BT123775-100GEA
36.08
USD
BT123775-100G
BT123775-500G
Tannic acid ACS
Tannic acid
CAS: 1401-55-4; EC No: 215-753-2; MDL No: MFCD00066397; RTECS: WW5075000
Powder; Molecular Formula: C76H52O46; MW: 1701.20
Melting Point: 218°; Flash point: 199°C (390°F)
- RTECS: WW5075000
- Powder
Formula:
C₇₆H₅₂O₄₆ MW: 1701.2 g/mol Melting Pt: 218 °C Flash Pt: 199 °C (390 °F) |
MDL Number:
MFCD00066397 CAS Number: 1401-55-4 EINECS: 215-753-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...