5-Bromosalicylaldehyde ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Bromo-2-hydroxybenzaldehyde

123760-100G 123760-25G 123760-500G
BT123760-100GEA 139 USD
BT123760-100G BT123760-25G BT123760-500G
5-Bromosalicylaldehyde ≥98%
5-Bromosalicylaldehyde

CAS: 1761-61-1; EC No: 217-167-2; MDL No: MFCD00003330 Powder; Molecular Formula: C7H5BrO2; MW: 201.02 Melting Point: 102-106° Air Sensitive


  • Air Sensitive
  • Powder

Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Melting Pt: 102…106 °C
MDL Number: MFCD00003330
CAS Number: 1761-61-1
EINECS: 217-167-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR