Methyl-4-aminosalicylate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Methyl 4-amino-2-hydroxybenzoate
123750-100G
123750-500G
BT123750-100GEA
222
USD
BT123750-100G
BT123750-500G
Methyl-4-aminosalicylate ≥97%
Methyl 4-aminosalicylate
CAS: 4136-97-4; EC No: 223-958-3; MDL No: MFCD00088091
Powder; Molecular Formula: C8H9NO3; MW: 167.18
- Powder
Formula:
C₈H₉NO₃ MW: 167.18 g/mol |
MDL Number:
MFCD00088091 CAS Number: 4136-97-4 EINECS: 223-958-3 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...