(±)-3-Hydroxybutyric acid, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
3-Hydroxybutyric acid, DL-3-Hydroxybutyric Acid, (±)-3-Hydroxybutanoic acid
123670-25G
123670-5G
BT123670-25GEA
234
USD
BT123670-25G
BT123670-5G
(±)-3-Hydroxybutyric acid, Technical Grade
(±)-3-Hydroxybutyric acid
CAS: 300-85-6; EC No: 206-099-9; MDL No: MFCD00004546
Liquid; Molecular Formula: C4H8O3; MW: 104.10
Boiling Point: 118-120°/2 mmHg; Flash point: 112°C (234°F)
Density (g/mL): 1.126; Refractive Index: 1.443
Hygroscopic
- Refractive index: 1.443
- Hygroscopic
- Liquid
Formula:
C₄H₈O₃ MW: 104.11 g/mol Boiling Pt: 118…120 °C/2 mmHg Density: 1.126 g/mL Flash Pt: 112 °C (234 °F) |
MDL Number:
MFCD00004546 CAS Number: 300-85-6 EINECS: 206-099-9 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...