D-(+)-Galactose ≥98%

Supplier: BeanTown Chemical

Synonyms: D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal

123650-1KG 123650-250G 123650-50G
BT123650-1KGEA 223.29 USD
BT123650-1KG BT123650-250G BT123650-50G
D-(+)-Galactose ≥98%
D-(+)-Galactose

CAS: 59-23-4; EC No: 200-416-4; MDL No: MFCD00151230; RTECS: LW5490000 Powder; Molecular Formula: C6H12O6; MW: 180.16 Melting Point: 168-170°


  • RTECS: LW5490000
  • Powder

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 168…170 °C
MDL Number: MFCD00151230
CAS Number: 59-23-4
EINECS: 200-416-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=508b1d21a6801ddfb1a5f13a64cb06e6f95e8922&vid=3c02003b1d12c9bfd11c2e6faf5cc37d4325d208&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR