1,2-Cyclopentane diformic anhydride

Supplier: BeanTown Chemical

Synonyms: Tetrahydro-1H-cyclopenta[c]furan-1,3(3ah)-dione

123630-1G 123630-5G
BT123630-1GEA 112 USD
BT123630-1G BT123630-5G
1,2-Cyclopentane diformic anhydride
1,2-Cyclopentane diformic anhydride

CAS: 5763-49-5; EC No: 227-286-1 Solid; Molecular Formula: C7H8O3; MW: 140.14


  • Solid

Formula: C₇H₈O₃
MW: 140.138 g/mol
CAS Number: 5763-49-5
EINECS: 227-286-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR