2-Deoxy-D(-)-ribose ≥99%

Supplier: BeanTown Chemical

123610-1G 123610-25G 123610-5G
BT123610-1GEA 21 USD
BT123610-1G BT123610-25G BT123610-5G
2-Deoxy-D(-)-ribose ≥99%
2-Deoxy-D(-)-ribose

CAS: 533-67-5; EC No: 208-573-0; MDL No: MFCD00135904; RTECS: SB7230000 Powder; Molecular Formula: C5H10O4; MW: 134.13 Melting Point: 89-90° Optical Rotation: [α]22/D -59°, c = 1 in water Hygroscopic


  • RTECS: SB7230000
  • Optical rotation: [α]22/D -59°, c = 1 in water
  • Hygroscopic
  • Powder

Formula: C₅H₁₀O₄
Melting Pt: 89…90 °C
MDL Number: MFCD00135904
CAS Number: 533-67-5
EINECS: 208-573-0

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR