6-Bromo-4-quinazolone ≥95%

Supplier: BeanTown Chemical

Synonyms: 6-Bromo-4(3H)-quinazolinone

123600-1G 123600-5G
BT123600-1GEA 49 USD
BT123600-1G BT123600-5G
6-Bromo-4-quinazolone ≥95%
6-Bromo-4-quinazolone

CAS: 32084-59-6 Solid; Molecular Formula: C8H5BrN2O; MW: 225.04


  • Solid

Formula: C₈H₅BrN₂O
MW: 225.04 g/mol
MDL Number: MFCD01219541
CAS Number: 32084-59-6

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR