Hafnium diboride ≥99.5% (trace metals basis excluding Zr)

Supplier: BeanTown Chemical

123595-10G 123595-250G 123595-50G
BT123595-10GEA 58 USD
BT123595-10G BT123595-250G BT123595-50G
Hafnium diboride ≥99.5% (trace metals basis excluding Zr)
Hafnium diboride

CAS: 12007-23-7; EC No: 234-500-7; MDL No: MFCD00016125 -325 Mesh Powder ; Linear Formula: HfB2; MW: 200.11 Melting Point: 3100° Density (g/mL): 10.5


  • -325 Mesh Powder

Formula: B₂Hf
MW: 200.11 g/mol
Melting Pt: 3100 °C
Density: 10.5 g/mL
MDL Number: MFCD00016125
CAS Number: 12007-23-7
EINECS: 234-500-7

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR