4-Methoxy-1-butanol ≥98%
Supplier: BeanTown Chemical
123555-1G
123555-5G
BT123555-1GEA
38
USD
BT123555-1G
BT123555-5G
4-Methoxy-1-butanol ≥98%
4-Methoxy-1-butanol
CAS: 111-32-0; EC No: 203-858-6; MDL No: MFCD00040437; RTECS: EL4550000
Liquid; Molecular Formula: C5H12O2 ; MW: 104.15
Boiling Point: 66°/7 mmHg; Flash point: 73°C (163°F)
Density (g/mL): 0.93
- RTECS: EL4550000
- Liquid
Formula:
C₅H₁₂O₂ Boiling Pt: 66 °C/7 mmHg Density: 0.93 g/mL Flash Pt: 73 °C (163 °F) |
MDL Number:
MFCD00040437 CAS Number: 111-32-0 EINECS: 203-858-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...