9-Anthracenemethanol ≥97%
Supplier: BeanTown Chemical
Synonyms:
9-Anthrylmethanol, Anthracene-9-methanol
123530-10G
123530-250G
123530-50G
BT123530-10GEA
61.78
USD
BT123530-10G
BT123530-250G
BT123530-50G
9-Anthracenemethanol ≥97%
9-Anthracenemethanol
CAS: 1468-95-7; EC No: 215-998-5; MDL No: MFCD00001264; RTECS: CB0577500
Solid; Molecular Formula: C15H12O; MW: 208.26
- RTECS: CB0577500
- Solid
Formula:
C₁₅H₁₂O MW: 208.26 g/mol |
MDL Number:
MFCD00001264 CAS Number: 1468-95-7 EINECS: 215-998-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...