Diboron trioxide ≥99%

Supplier: BeanTown Chemical

123525-2.5KG 123525-500G
BT123525-2.5KGEA 370.51 USD
BT123525-2.5KG BT123525-500G
Diboron trioxide ≥99%
Diboron trioxide

CAS: 1303-86-2; EC No: 215-125-8; MDL No: MFCD00011315; RTECS: ED7900000 Crystalline; Molecular Formula: B2O3; MW: 69.62 Melting Point: 45° Density (g/mL): 2.46 Hygroscopic


  • RTECS: ED7900000
  • Hygroscopic
  • Crystalline

Formula: B₂O₃
Melting Pt: 45 °C
Density: 2.46 g/mL
MDL Number: MFCD00011315
CAS Number: 1303-86-2
EINECS: 215-125-8

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR