7-Chloro-1H-pyrrolo[2,3-c]pyridine ≥97%
Supplier: BeanTown Chemical
Synonyms:
7-Chloro-6-azaindole
123465-1G
123465-250MG
123465-5G
BT123465-1GEA
188
USD
BT123465-1G
BT123465-250MG
BT123465-5G
7-Chloro-1H-pyrrolo[2,3-c]pyridine ≥97%
7-Chloro-1H-pyrrolo[2,3-c]pyridine
CAS: 357263-41-3; MDL No: MFCD03839905
Solid; Molecular Formula: C7H5ClN2; MW: 152.58
- Solid
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol |
MDL Number:
MFCD03839905 CAS Number: 357263-41-3 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...