2,5-Dibromo-4-picoline ≥97%
Supplier: BeanTown Chemical
Synonyms:
2,5-Dibromo-4-methylpyridine
123410-1G
123410-5G
BT123410-1GEA
82
USD
BT123410-1G
BT123410-5G
2,5-Dibromo-4-picoline ≥97%
2,5-Dibromo-4-picoline
CAS: 3430-26-0; MDL No: MFCD00234955
Low Melting Solid; Molecular Formula: C6H5Br2N; MW: 250.92
Melting Point: 37-42°
- Low Melting Solid
Formula:
C₆H₅Br₂N MW: 250.92 Melting Pt: 37…42 °C |
MDL Number:
MFCD00234955 CAS Number: 3430-26-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...