Yttrium(III) 2-ethylhexanoate ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
123265-100G
123265-25G
123265-5G
BT123265-100GEA
466
USD
BT123265-100G
BT123265-25G
BT123265-5G
Yttrium(III) 2-ethylhexanoate ≥99.5% (trace metals basis)
Yttrium (III) 2-ethylhexanoate
CAS: 103470-68-4; MDL No: MFCD00070441
Powder; Molecular Formula: C24H45O6Y ; MW: 518.16
- Powder
Formula:
C₂₄H₄₅O₆Y MW: 518.524 g/mol |
MDL Number:
MFCD00070441 CAS Number: 103470-68-4 |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...