Sulfanilamide ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Aminobenzenesulphonamide

123260-100G 123260-2.5KG 123260-500G
BT123260-100GEA 32.86 USD
BT123260-100G BT123260-2.5KG BT123260-500G
Sulfanilamide ≥98%
Sulphanilamide

CAS: 63-74-1; EC No: 200-563-4; MDL No: MFCD00007939; RTECS: WO8400000 Solid; Linear Formula: H2NC6H4SO2NH2; Molecular Formula: C6H8N2O2S; MW: 172.21 Melting Point: 163-166° Density (g/mL): 1.08


  • RTECS: WO8400000
  • Solid

Formula: H₂NC₆H₄SO₂NH₂
MW: 172.21 g/mol
Melting Pt: 163…166 °C
Density: 1.08 g/mL
MDL Number: MFCD00007939
CAS Number: 63-74-1
EINECS: 200-563-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a76bb9ab6bdf23e9d846b24d4f57047dea1d5071&vid=f2f1c5e47a127cd33d3b4a2a31b1057c375456eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR