Magnesium niobium oxide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

123225-10G 123225-250G 123225-50G
BT123225-10GEA 48 USD
BT123225-10G BT123225-250G BT123225-50G
Magnesium niobium oxide ≥99.9% (trace metals basis)
Magnesium niobium oxide

CAS: 12163-26-7; EC No: 235-310-7; MDL No: MFCD00210631 -200 Mesh Powder; Molecular Formula: MgNb2O6 ; MW: 306.11


  • -200 Mesh Powder

Formula: MgNb₂O₆
MW: 306.11 g/mol
MDL Number: MFCD00210631
CAS Number: 12163-26-7
EINECS: 235-310-7

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR